Skip to Content
Merck
All Photos(1)

Documents

PHL89505

Galangin

phyproof® Reference Substance

Synonym(s):

3,5,7-Trihydroxyflavone, Norizalpinin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
272179
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

214-215 °C (lit.)

application(s)

cannabis testing

format

neat

SMILES string

Oc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3ccccc3

InChI

1S/C15H10O5/c16-9-6-10(17)12-11(7-9)20-15(14(19)13(12)18)8-4-2-1-3-5-8/h1-7,16-17,19H

InChI key

VCCRNZQBSJXYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.
Galangin, a polyphenol-based flavonol, belongs to the family of flavonoid compounds. It is abundantly available in human dietary sources such as fruits, vegetables, etc. and is widely used in herbal medicines, owing to its remedial properties.

Application

Galangin may be used as an analytical reference standard for the determination of the analyte in raw propolis collected by honeybees (Apis mellifica L.) and in the extracts of grape wine by chromatographic and colorimetric techniques.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galangin expresses bactericidal activity against multiple-resistant bacteria: MRSA, Enterococcus spp. and Pseudomonas aeruginosa
Pepeljnjak S and Kosalec I
FEMS Microbiology Letters, 240(1), 111-116 (2004)
Determination of flavonoids by high-performance liquid chromatography and capillary electrophoresis
Wang SP and Huang KJ
Journal of Chromatography A, 1032(1-2), 273-279 (2004)
Differential systemic exposure to galangin after oral and intravenous administration to rats
Chen F, et al.
Chemistry Central Journal, 9(1), 14-14 (2015)
Estimation of total flavonoid content in propolis by two complementary colorimetric methods
Chang CC, et al.
Journal of food and drug analysis, 10(3), 273-279 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service