Skip to Content
Merck
All Photos(1)

Key Documents

PHL89225

Hyperforin (stable Dicyclohexylammonium salt)

phyproof® Reference Substance

Synonym(s):

Hyperforin (dicyclohexylammonium) salt, Hyp-DCHA, Hyperforin-DCHA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,230.00

$1,230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C35H52O4·C12H23N
CAS Number:
Molecular Weight:
718.10
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)C(=O)[C@@]34C(O)=C(C\C=C(\C)C)C(=O)[C@@](C\C=C(\C)C)(C[C@H](C\C=C(/C)C)[C@@]3(C)CC\C=C(\C)C)C4=O

InChI

1S/C35H52O4.C12H23N/c1-22(2)13-12-19-33(11)27(16-14-23(3)4)21-34(20-18-25(7)8)30(37)28(17-15-24(5)6)31(38)35(33,32(34)39)29(36)26(9)10;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h13-15,18,26-27,38H,12,16-17,19-21H2,1-11H3;11-13H,1-10H2/t27-,33+,34+,35-;/m0./s1

InChI key

ZUYCAROZOLUHJY-DNSWOBEMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort.
Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found it to have neuroprotective effects against Alzheimer′s disease (AD) neuropathology, including the ability to disassemble amyloid-β aggregates in vitro and improve spatial memory in vivo.
Stable salt of hyperforin.
Stable salt of hyperforin. Hyperforin is believed to be the primary active constituent responsible for the antidepressant and anxiolytic properties of the extracts of St. John′s wort. It acts as a reuptake inhibitor of monoamines, including serotonin, norepinephrine, dopamine, and of GABA and glutamate by activating the transient receptor potential ion channel TRPC6. Activation of TRPC6 induces the entry of sodium and calcium into the cell which causes inhibition of monoamine reuptake. It also is an activator of the pregnane X receptor (PXR), which serves as a key regulator of CYP3A4 transcription, a member of the cytochrome (CYP) P450 enzyme system. Recent studies have found other neurological effects, effects on inflammation, as well as antibacterial, antitumoral and antiangiogenic effects.

Other Notes

This compound is commonly found in plants of the genus: hypericum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengkang He et al.
Clinical and translational medicine, 10(6), e205-e205 (2020-11-03)
Recurrent moderate hypoglycemia (RH), a major adverse effect of hypoglycemic therapy in diabetic patients, is one of the main risk factors for cognitive impairment and dementia. Transient receptor potential canonical channel 6 (TRPC6) is a potential therapeutic target for Alzheimer's

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service