Skip to Content
Merck
All Photos(1)

Key Documents

PHL83317

Isobavachalcone

phyproof® Reference Substance

Synonym(s):

(2E)-1-[2,4-Dihydroxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)-2-propen-1-one, 2′,4,4′-Trihydroxy-3′-(3-methyl-2-butenyl)-chalcone, Corylifolinin, Isobacachalcone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$546.00

$546.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$546.00

About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$546.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(CC=C(C)C)C(O)=C(C(/C=C/C2=CC=C(O)C=C2)=O)C=C1

InChI

1S/C20H20O4/c1-13(2)3-9-16-19(23)12-10-17(20(16)24)18(22)11-6-14-4-7-15(21)8-5-14/h3-8,10-12,21,23-24H,9H2,1-2H3/b11-6+

InChI key

DUWPGRAKHMEPCM-IZZDOVSWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Isobavachalcone is a chalcone isolated from multipurpose medical plant Psoralea corylifolia that inhibits LPS-induced ICAM-1 expression and leukocyte adhesion to brain endothelial cells. It appears that isobavachalcone modulates both MyD88-dependent and TRIF-dependent signaling of toll-like receptor 4 (TLR4). Isobavachalcone significantly inhibits both oligomerization and fibrillization of Aβ42. Isobavachalcone exhibits a wide spectrum of biological activities including antioxidative, antiplatelet, antimicrobial, anti-inflammatory, antitumor, and neuroprotective.
Isobavachalcone is a chalcone isolated from multipurpose medical plant Psoralea corylifolia that inhibits LPS-induced ICAM-1 expression and leukocyte adhesion to brain endothelial cells.

Other Notes

This compound is commonly found in plants of the genus: angelica glycyrrhiza

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service