Skip to Content
Merck
All Photos(1)

Key Documents

PHL80892

Atropine sulfate

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$507.00

$507.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$507.00

About This Item

Empirical Formula (Hill Notation):
C17H23NO3 · 0.5H2SO4
CAS Number:
Molecular Weight:
338.41
UNSPSC Code:
41116107
NACRES:
NA.24

$507.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

[S](=O)(=O)([O-])[O-].[N+H]4(C5CCC4CC(C5)OC(=O)C(CO)c6ccccc6)C.[N+H]1(C2CCC1CC(C2)OC(=O)C(CO)c3ccccc3)C

InChI

1S/2C17H23NO3.H2O4S/c2*1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12;1-5(2,3)4/h2*2-6,13-16,19H,7-11H2,1H3;(H2,1,2,3,4)

InChI key

HOBWAPHTEJGALG-UHFFFAOYSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

For the analysis of the phytotoxins. Tropane alkaloid.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Bohnert et al.
Toxicology letters, 324, 86-94 (2020-01-20)
Organophosphorus nerve agents (NA) inhibit acetylcholinesterase (AChE) which results in the over-stimulation of both the central and peripheral nervous systems, creating a toxic syndrome that can be lethal if left untreated (Cannard, 2006). It is standard practice to treat Sarin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service