Skip to Content
Merck
All Photos(1)

Key Documents

P6349

Sigma-Aldrich

TWEEN® 80

viscous liquid, Low-peroxide

Synonym(s):

POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$1,370.00

$1,370.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$1,370.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

$1,370.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

description

non-ionic

form

viscous liquid

mol wt

average mol wt ~1310

contains

BHT as antioxidant

composition

Oleic acid, ~70% (balance primarily linoleic, palmitic, and stearic acids)

impurities

≤0.5 μmol/g Peroxides
≤1.0 μmol/g Carbonyls
≤3.0% water

density

1.07 g/cm3 at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rokeya Tasneen et al.
Antimicrobial agents and chemotherapy, 52(10), 3664-3668 (2008-08-13)
PA-824 is in phase II clinical testing to treat tuberculosis. At a dose of 100 mg/kg of body weight, it has demonstrated bactericidal activity during the initial and continuation phases of treatment in a murine model of tuberculosis. In a
Syed A Sattar et al.
PloS one, 6(2), e17340-e17340 (2011-03-11)
Human noroviruses (HuNoV), a major cause of acute gastroenteritis worldwide, cannot be readily cultured in the lab. Therefore, a feline calicivirus (FCV) is often used as its surrogate to, among other things, test alcohol-based handrubs (ABHR). The more recent laboratory
Cristina Marcos-Arias et al.
BMC complementary and alternative medicine, 11, 119-119 (2011-11-29)
Candida-associated denture stomatitis is a frequent infectious disease. Treatment of this oral condition is difficult because failures and recurrences are common. The aim of this study was to test the in vitro antifungal activity of pure constituents of essentials oils.
Hau-Ren Chen et al.
International journal of molecular sciences, 12(11), 8245-8258 (2011-12-17)
The separation of mercury ions from artificially contaminated water by the foam fractionation process using a biosurfactant (surfactin) and chemical surfactants (SDS and Tween-80) was investigated in this study. Parameters such as surfactant and mercury concentration, pH, foam volume, and
Qun Wang et al.
Brain research, 1285, 188-195 (2009-06-17)
The group IIA secretory phospholipase A2 (sPLA(2)-IIA) has been studied extensively because of its involvement in inflammatory processes. Up-regulation of this enzyme has been shown in a number of neurodegenerative diseases including cerebral ischemia and Alzheimer's disease. PX-18 is a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service