Skip to Content
Merck
All Photos(1)

Documents

P0914

Sigma-Aldrich

Protein standard

Micro Standard, liquid

Synonym(s):

Albumin from bovine serum, protein standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

form

liquid

contains

0.05% sodium azide as preservative

packaging

ampule of 1 mL

technique(s)

ELISA: suitable

color

clear colorless

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Application

Concentrated solution for all protein assays.
Protein standard has been used:

  • as a standard in the quantification of protein release by bicinchoninic acid assay
  • as a standard in the quantification of urea by Bradfordassay
  • as a molecular size marker for the quantification of protein by native polyacrylamide gel electrophoresis
Protein standard for all protein assays. The sealed ampules provide assurance that the concentration will be correct with every assay.

Packaging

Sealed ampules

Other Notes

1 mg bovine serum albumin/ml in 0.15 M NaCl

Suitability

This product is commonly used for protein determination by the Bicinchoninic Acid (BCA) method.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alexander Pekarsky et al.
Bioprocess and biosystems engineering, 42(10), 1611-1624 (2019-07-04)
In biotechnological processes, technical failures in the upstream process often lead to batch loss. It is of great interest to investigate the empirical impact of technical failures to understand and mitigate their impact accurately and reduce economic damage. We investigated
Screening and characterization of a novel esterase from a metagenomic library
Yun-Jung K, et al.
Protein Expression and Purification, 45(2), 315-323 (2006)
Alexander Pekarsky et al.
Bioprocess and biosystems engineering, 44(6), 1049-1061 (2021-01-26)
Technical failures lead to deviations in process parameters that can exceed studied process boundaries. The impact on cell and target protein is often unknown. However, investigations on common technical failures might yield interesting insights into process and protein robustness. Recently
Mudalige Don Hiranya Jayasanka Senavirathna et al.
Water environment research : a research publication of the Water Environment Federation, 95(1), e10834-e10834 (2023-01-13)
This study demonstrated the temporal variation of 2-methylisoborneol (2-MIB) and geosmin (GSM) production of two filamentous cyanobacteria species Pseudanabaena galeata (NIES-512; planktonic) and Phormidium ambiguum (NIES-2119; benthic) exposed to high light intensity (950-1000 μmol m-2  s-1 photosynthetically active radiation). The production of
Shouliang Xiong et al.
Bioengineered, 12(2), 11138-11155 (2021-11-17)
Osteoarthritis (OA) is characterized by destruction of articular cartilage with an imbalance between synthesis and degradation of extracellular matrix (ECM). In the current study, we explored the role of microRNA-34a (miR-34a) and the behind epigenetic mechanism in the degradation of

Articles

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Protocols

To standardize a procedure for the determination of protein by modified Lowry.

To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.

Related Content

Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service