Skip to Content
Merck
All Photos(1)

Key Documents

O0380

Supelco

Oxymetholone

analytical standard

Synonym(s):

17β-Hydroxy-2-(hydroxymethylene)-17-methyl-5α-androstan-3-one, Anasterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95%

form

solid

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to light yellow

solubility

H2O: ≤0.5 mg/mL
ethanol: 6.7 mg/mL

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@]3([H])C(CC[C@@]4(C)[C@@]3([H])CC[C@]4(C)O)[C@@]1(C)C\C(=C\O)C(=O)C2

InChI

1S/C21H32O3/c1-19-11-13(12-22)18(23)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,22,24H,4-11H2,1-3H3/b13-12-/t14-,15+,16?,17-,19-,20-,21-/m0/s1

InChI key

ICMWWNHDUZJFDW-ZDBQEZQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Androgen

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cortney L Ballard et al.
Behavioral neuroscience, 119(3), 752-758 (2005-07-07)
Previous studies by the authors have shown voluntary intracerebroventricular (icv) testosterone self-administration in hamsters (Mesocricetus auratus). Here, the authors compared icv self-administration of 4 anabolic steroids (drostanolone, nandrolone, oxymetholone, and stanozolol) at 0.1, 1.0, and 2.0 microg/microl, each for 8
Zerrin Yilmaz et al.
Journal of pediatric hematology/oncology, 27(10), 565-566 (2005-10-12)
A 13-year-old girl with a history of Fanconi anemia developed acute myeloid leukemia of the M7 subtype with a 45,XX,-7 karyotype, which is rare in M7 subtype. Treatment protocols were set up, but she died of sepsis and osteomyelitis during
[Peliosis hepatitis secondary to hormone treatment].
M Tallón García et al.
Anales de pediatria (Barcelona, Spain : 2003), 75(4), 286-288 (2011-07-08)
[Bone marrow failure due to telomere associated gene mutation].
Hiroki Yamaguchi et al.
[Rinsho ketsueki] The Japanese journal of clinical hematology, 51(8), 646-653 (2010-09-02)
Gordon C Hard et al.
Toxicologic pathology, 40(3), 473-481 (2012-02-03)
From the archives of the National Toxicology Program, National Institutes of Health, kidney sections from twenty-four carcinogenicity studies (representing twenty-three chemicals) in male and female F344 rats were histopathologically re-evaluated to grade the severity of chronic progressive nephropathy (CPN) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service