Skip to Content
Merck
All Photos(2)

Documents

NIST84L

Potassium hydrogen phthalate

NIST® SRM® 84l

Synonym(s):

Potassium phthalate monobasic, KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

packaging

pkg of 60 g

manufacturer/tradename

NIST®

technique(s)

titration: suitable

mp

295-300 °C (dec.) (lit.)

format

matrix material

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

It consists of highly purified potassium hydrogen phthalate (KHP), KHC8H4O4. Its certified value is calculated as the mass fraction of total acid (replaceable H+) expressed as KHP. Its certified value is based on the results of coulometric assays of ground and dried material. The assay value for this material was obtained by automated coulometric titration to the inflection point (pH ≈ 8.4) of weighed test portions of KHP. The certified value represents the result of 30 titrations of test portions taken from 16 bottles selected by stratified random sampling.

The certification of SRM 84L is valid, within the measurement uncertainties specified, provided the RM is handled and stored in accordance with the instructions given in the “Instructions for Storage and Use”.

SRM 84L_CERT

SRM 84L_SDS

Application

This Standard Reference Material (SRM) is intended for use as an acidimetric primary standard. This SRM is certified for acidimetric assay ONLY and is not intendedfor use as a pH standard.

Features and Benefits

  • A unit of SRM 84L is supplied as crystalline material in a 60 g unit
  • Information on product description, disposal, and transportation is available in the accompanying MSDS
  • The enclosed CoA contains information on the SRM’s expiry date and instructions for its handling, storage, and use

Other Notes

Certified for the analytes listed below.
Potassium Hydrogen Phthalate

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service