Skip to Content
Merck
All Photos(1)

Documents

NIST8404

Almond Flour for allergen detection

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 5x5 mg

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

General description

One unit of almond Flour, RM 8404, contains five packets of blanched almond flour packets, each containing ~5 grams of material.
For more information, please refer to the SDS and COA.
SRM 8404_cert
SRM 8404_SDS

Application
RM 8404 is intended to harmonize the measurement of allergenic almond proteins in foods, considering the following:
(1) the composition of almonds may vary depending on their source or the pretreatments they have undergone (e.g., roasting, defatting)
(2) when the material undergoes preparation or processing methods (such as boiling, frying, or baking), its composition and properties may be altered.

Features and Benefits

  • Non-certified mass fraction values are provided forproximates on an as received basis and values are metrologically traceable tothe materials and methods used to determine it.
  • Thenon-certified values are ideal for method harmonization, method development,and process control.
  • Expirationdetail, homogeneity assessment, instructions for storage and uses are providedon the NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Protein, Fat, Solids, Ash, Calories

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service