Skip to Content
Merck
All Photos(1)

Key Documents

NIST8044

Common Commercial Asbestos: Chrysotile

NIST®SRM®

Synonym(s):

Chrysotile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg3Si2H4O9
CAS Number:
Molecular Weight:
277.11
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

packaging

pkg of 2.6 g

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

application(s)

industrial hygiene

format

matrix material

SMILES string

[Si]([O-])([O-])([O-])O.[Si]([O-])([O-])([O-])O.[Mg+2].[Mg+2].[Mg+2].O

InChI

1S/3Mg.2HO4Si.H2O/c;;;2*1-5(2,3)4;/h;;;2*1H;1H2/q3*+2;2*-3;

InChI key

CWBIFDGMOSWLRQ-UHFFFAOYSA-N

General description

One unit of RM 8044 comprises a single bottle of loosely packed mine-grade chrysotile asbestos, with each bottle containing around 2.6 grams of material. For more information, please refer to the COA and SDS.SRM 8044_cert

SRM 8044_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service