Skip to Content
Merck
All Photos(1)

Key Documents

NIST46H

Portland cement

NIST® SRM® 46h, fineness standard

Synonym(s):

Portland Cement Fineness Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 10 vials x 5 g

manufacturer/tradename

NIST®

Looking for similar products? Visit Product Comparison Guide

General description

A unit of Standard Reference Material (SRM) 46h contains 10 glass vials of approximately 5 g of powdered cement. Each vial has a plastic snap off cap and is contained in a sealed foil bag.

SRM 46H_cert

SRM 46H_SDS

Application

This SRM is intended for use in calibrating fineness testing equipment according to ASTM Standard Methods.

Features and Benefits

  • Values for constituents are listed in the NIST certificate of analysis.
  • The health and safety information are provided in the safety data sheet.

Other Notes

Reference/Informational value is provided for the following analytes.
Calcium Oxide (CaO), Silicon Dioxide (SiO2), Aluminum Oxide (Al2O3), Ferric Oxide (Fe2O3), Sulfur Trioxide (SO3), Potassium Oxide (K2O), Titanium Dioxide (TiO2), Phosphorus Pentoxide (P2O5), Sodium Oxide (Na2O), Magnesium Oxide (MgO), Tricalcium Silicate (C3S), Dicalcium Silicate (C2S), Tricalcium Aluminate (C3A), Tetracalcium Alumino-ferrite (C4AF)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service