Skip to Content
Merck
All Photos(1)

Key Documents

NIST3136

Nickel (Ni) standard solution

NIST® SRM® 3136

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

One unit of SRM 3136 comprises five sealed borosilicate glass ampoules, each containing 10 mL of an acidified aqueous solution. This solution is gravimetrically prepared to contain a known mass fraction of nickel and comprised of nitric acid at an approximate volume fraction of ~ 8 %.


SRM 3136_cert

SRM 3136_SDS

Application

The SRM is intended for use as a primary calibration standard for the determination of nickel quantitatively.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values of Nickel, which is obtained through two methods:
  1. Gravimetric preparation
  2. Inductively coupled plasma optical emission spectrometry (ICP-OES).
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as milligram per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nickel (Ni)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service