Skip to Content
Merck
All Photos(1)

Documents

NIST3074

Phthalates in methanol

NIST® SRM® 3074

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

format

multi-component solution

General description

This Standard Reference Material (SRM) is a solution of one adipate and six phthalates in methanol intended primarily for use in the calibration of chromatographic instrumentation used for the determination of the certified components in the mixture. Because of its miscibility with water, this SRM can also be used to fortify samples with known amounts of the six phthalates. A unit of SRM 3074 consists of five 2 mL ampoules, each containing 1.2 mLof solution. For more information, please refer ot the COA and SDS.

SRM 3074_cert

SRM 3074_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Dimethylphthalate, Dimethylphthalate, Di-n-butylphthalate, Benzylbutylphthalate, Bis(2-ethylhexyl)phthalate, Di-n-octylphthalate

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service