Skip to Content
Merck
All Photos(1)

Documents

NIST2968

3-EPI-25-HYDROXYVITAMIN D3 CALIBRATION SOLUTION

NIST®SRM®

Synonym(s):

3-EPI-25-HYDROXYVITAMIN D3 CALIBRATION SOLUTION

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H44O2
UNSPSC Code:
41116107

format

mixture

storage temp.

−20°C

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service