Skip to Content
Merck
All Photos(1)

Key Documents

NIST2613A

Carbon monoxide in air

NIST® SRM® 2613a, nominal 20 μmol/mol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of cylinder

manufacturer/tradename

NIST®

application(s)

air monitoring
environmental

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

his Standard Reference Material (SRM) is a primary gas mixture for which the amount-of-substance fraction, expressed as concentration, may be related to secondary working standards. This SRM is intended for the calibration of instruments used for carbon monoxide determinations and for other applications. For more information, please refer to the SDS and COA.

SRM 2613a_cert SRM 2613a _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service