Skip to Content
Merck
All Photos(1)

Documents

NIST2373

Genomic DNA Standards for HER2 Measurements

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

liquid

manufacturer/tradename

NIST®

technique(s)

PCR: suitable
qPCR: suitable

application(s)

genomic analysis

storage temp.

2-8°C

General description

Genomic DNA Standards are standard reference materials (SRM 2373) containing genomic DNA extracted from five breast cancer cell lines with different amounts of amplification of the HER2 gene. The five components are genomic DNA materials derived from human cell lines SK-BR-3, MDA-MB-231, MDA-MB-361, MDA-MB-453, and BT-474, labeled A, B, C, D, and E, respectively. A unit of SRM 2373 consists of five vials, one of each component, containing approximately 100 μL of DNA solution.

SRM_2373_cert

SRM_2373_SDS

Application

Genomic DNA Standards for HER2 Measurements is intended for use in the assignment of ratios of the human epidermal growth factor receptor 2 gene (HER2, the official gene symbol ERBB2) to unamplified reference genes. It is suitable for use with quantitative polymerase chain reaction (qPCR) and digital polymerase chain reaction (dPCR) assays.

Features and Benefits

The use of this material will provide confidence in the assignment of certified ratios to the secondary standard DNA solutions for routine use with qPCR and dPCR assays.

Other Notes

  • SRM 2373 is intended for research use. This is a human source material.
  • Expiration details, storage, and usage instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua-Jun He et al.
Clinical chemistry and laboratory medicine, 57(8), 1142-1152 (2019-05-22)
Background The National Institute of Standards and Technology (NIST) Reference Material RM 8366 was developed to improve the quality of gene copy measurements of EGFR (epidermal growth factor receptor) and MET (proto-oncogene, receptor tyrosine kinase), important targets for cancer diagnostics
Hua-Jun He et al.
Biomolecular detection and quantification, 8, 1-8 (2016-06-24)
NIST standard reference material (SRM) 2373 was developed to improve the measurements of the HER2 gene amplification in DNA samples. SRM 2373 consists of genomic DNA extracted from five breast cancer cell lines with different amounts of amplification of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service