Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 150 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2168_cert
SRM 2168_SDS
Other Notes
Sulfur (S)
Reference/Informational value is provided for the following analytes.
Boron (B), Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Manganese (Mn), Nickel (Ni), Nitrogen (N), Oxygen (O), Phosphorus (P), Aluminum (Al), Antimony (Sb), Arsenic (As), Bismuth (Bi), Cadmium (Cd), Calcium (Ca), Lead (Pb), Magnesium (Mg), Molybdenum (Mo), Niobium (Nb), Selenium (Se), Silicon (Si), Tantalum (Ta), Tellurium (Te), Tin (Sn), Titanium (Ti), Tungsten (W), Vanadium (V), Zinc (Zn), Zirconium (Zr)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service