Skip to Content
Merck
All Photos(2)

Key Documents

NIST189C

Potassium tetraoxalate dihydrate

NIST® SRM® 189c, pH standard

Synonym(s):

Oxalic acid hemipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH3(C2O4)2 · 2H2O
CAS Number:
Molecular Weight:
254.19
Beilstein:
3854419
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product NIST189C is currently not available for purchase. Contact Technical Service

grade

certified reference material

Quality Level

form

crystals

packaging

pkg of 65 g

manufacturer/tradename

NIST®

application(s)

general analytical

SMILES string

O.O.[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

InChI

1S/2C2H2O4.K.2H2O/c2*3-1(4)2(5)6;;;/h2*(H,3,4)(H,5,6);;2*1H2/q;;+1;;/p-1

InChI key

KPFHSTZUXXJAES-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service