Skip to Content
Merck
All Photos(1)

Key Documents

NIST1412A

Multicomponent Glass (disk form)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

disc

packaging

pkg of 2 x9.5 g

manufacturer/tradename

NIST®

application(s)

glass & ceramic

storage temp.

15-25°C

General description

A unit of SRM 1412a contains two disks, each measuring approximately 40 mm in diameter, 3 mm in thickness, and weighing ~ 9.5 g. For more information, please refer to the COA and SDS.

SRM 1412A_cert


SRM 1412A_SDS

Application

The Standard Reference Material (SRM) is primarily intended for validating chemical and instrumental methods used to analyze the elemental composition of glass. It is also used for validating the value assignment of a laboratory′s in-house reference materials.

Features and Benefits

  • NIST-certified mass fraction values for elements in glass matrix are provided. The values are metrologically traceable to the SI unit of mass fraction and expressed as percent.
  • Information values for various constituents in multicomponent glass are provided as mass fraction (%).
  • Maintenance and expiration of SRM certification, instructions for handling, storage, and use are provided on NIST certificate

Other Notes

Certified for the analytes listed below.
Aluminum (Al) , Antimony (Sb), Arsenic (As), Barium (Ba), Boron (B), Calcium (Ca), Chromium (Cr), Iron (Fe), Potassium (K), Lead (Pb), Lithium (Li), Magnesium (Mg), Silicon (Si), Silver (Ag), Sodium (Na), Strontium (Sr), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Gallium (Ga), Phosphorus (P), Selenium (Se), Sulfur (S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service