Skip to Content
Merck
All Photos(1)

Key Documents

NIST1017B

Glass (particle size)

NIST® SRM® 1017b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

beads

packaging

pkg of 70 g

manufacturer/tradename

NIST®

application(s)

glass & ceramic

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1017b contains a glass bottle measuring ~70g of solid spherical soda-lime glass spheres.

SRM 1017B_cert

SRM 1017B_SDS

Application

The Standard Reference Material (SRM) is primarily intended for the evaluation and calibration of particle size measuring instruments within the 100 μm to 400 μm range. It is also utilized for evaluating wire-cloth test sieves that range from 106 μm (No. 140) to 355 μm (No. 45).

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values, which were obtained through two methods:
  1. scanning electron microscopy (SEM)
  2. standard sieving procedures on stratified and randomly selected bottles of spheres.
  • NIST certificate is provided with information values for nominal and effective sieve openings.
  • The certified values are metrologically traceable to the SI derived unit for mass fraction (expressed as percent).
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service