Skip to Content
Merck
All Photos(1)

Documents

N1230000

Norfloxacin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-piperazino-3-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18FN3O3
CAS Number:
Molecular Weight:
319.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

norfloxacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1C=C(C(O)=O)C(=O)c2cc(F)c(cc12)N3CCNCC3

InChI

1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23)

InChI key

OGJPXUAPXNRGGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norfloxacin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Norfloxacin blocks DNA replication by interfering with an ATP-induced structural transition of DNA complexed with DNA gyrase (topoisomerase).
Mode of action: inhibits bacterial DNA replication
Antimicrobial spectrum: Gram-negative bacteria; less effective against Gram-positive bacteria

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Nour El-Houda Jlili et al.
Journal of medical microbiology, 63(Pt 2), 195-202 (2013-11-07)
The prevalence of plasmid-mediated quinolone resistance genes [qnr, aac(6')-Ib-cr and qepA] was sought among Enterobacteriaceae strains obtained from the Children's Hospital of Tunis (Tunisia). Non-duplicate isolates (n = 278) with resistance to extended-spectrum cephalosporins and collected in 2003, 2007, 2008 and 2009
G E Stein
The American journal of medicine, 82(6B), 18-21 (1987-06-26)
The pharmacokinetic profile of norfloxacin, an oral fluoroquinolone, is more complex than that of many antibacterial agents. Following administration of a 400-mg dose, peak serum concentrations of 1.5 to 2.0 micrograms/ml are achieved within one to two hours. The drug
L Miano et al.
European urology, 17 Suppl 1, 13-18 (1990-01-01)
A review of worldwide clinical trials with norfloxacin in the treatment of uncomplicated urinary tract infections (UTIs), as well as our personal experience with 215 assessable patients, is presented. Almost all patients received 400 mg b.i.d. for 3-15 days. Bacteriological
Bacterial resistance to norfloxacin and other newer quinolones: pattern and prospects.
D R Nalin
European journal of cancer & clinical oncology, 24 Suppl 1, S55-S61 (1988-01-01)
E J Goldstein
The American journal of medicine, 82(6B), 3-17 (1987-06-26)
Norfloxacin is an orally absorbed fluoroquinolone antibacterial with a fluorine at position 6 and a piperazine ring at position 7. These changes have resulted in a marked enhancement (compared with that of the older quinolones) of in vitro antibacterial activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service