Skip to Content
Merck
All Photos(1)

Key Documents

M6907

Millipore

Malt Extract Agar

Microbiologically tested, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$133.00

$133.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$133.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

$133.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Quality Level

sterility

non-sterile

form

powder

manufacturer/tradename

NutriSelect® Plus

application(s)

bioburden testing
environmental
food and beverages
pharmaceutical
sterility testing

suitability

bacteria

General description

Isolation and detection of yeasts and molds

Application

Malt Extract Agar has been used in:
  • isolation of ascomycetes from orchid roots[1]
  • cultivation of Pleurotus ferulae mycelium[2]
  • the culturing of fungal species[3]

Other Notes

Contains maltose, dextrin, glycerol, peptone and agar

Preparation Note

Solve 33.6 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orchids keep the ascomycetes outside: a highly diverse group of ascomycetes colonizing the velamen of epiphytic orchids from a tropical mountain rainforest in Southern Ecuador
Herrera P, et al.
Mycology, 1(4), 262-268 (2010)
Antioxidative Potential, Antibiofilm Activity and Partial Chemical Characterization of Cultured Broth after Pleurotus ferulae Mycelium Cultivation
Vamanu E, et al.
Reviews in Fluorescence, 69(11), 2996-3000 (2018)
Michael J Dillon et al.
Microbial biotechnology, 10(2), 381-394 (2016-12-21)
The fungus Batrachochytrium dendrobatidis (Bd) causes chytridiomycosis, a lethal epizootic disease of amphibians. Rapid identification of the pathogen and biosecurity is essential to prevent its spread, but current laboratory-based tests are time-consuming and require specialist equipment. Here, we describe the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service