Skip to Content
Merck
All Photos(1)

Key Documents

M5029

Supelco

(±)-3,4-Methylenedioxymethamphetamine hydrochloride solution

analytical standard, for drug analysis

Synonym(s):

DL-3,4-Methylenedioxymethamphetamine hydrochloride solution, DL-MDMA hydrochloride solution, XTC hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2 · HCl
CAS Number:
Molecular Weight:
229.70
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CNC(C)Cc1ccc2OCOc2c1

InChI

1S/C11H15NO2.ClH/c1-8(12-2)5-9-3-4-10-11(6-9)14-7-13-10;/h3-4,6,8,12H,5,7H2,1-2H3;1H

InChI key

LUWHVONVCYWRMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Methylenedioxymethamphetamine is considered as an illicit drug. It stimulates neurons to release serotonin and dopamine.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-substituted piperazines abused by humans mimic the molecular mechanism of 3,4-methylenedioxymethamphetamine (MDMA, or 'Ecstasy').
Baumann MH
Neuropsychopharmacology, 30(3), 550-560 (2005)
Nicola C Newton et al.
Journal of psychoactive drugs, 44(5), 372-380 (2013-03-06)
The Climate Schools: Ecstasy module is a universal harm-minimisation school-based prevention program for adolescents aged 14 to 16 years. The program was developed to address the need for Ecstasy prevention given the increasing use of Ecstasy use among young Australians.
Céline Eiden et al.
Journal of psychoactive drugs, 45(1), 94-97 (2013-05-11)
Typical scenarios of drug-facilitated sexual assaults usually involve victims having ingested a drink after which they had little, partial or no recollection of events for a period of time. We were surprised by the case of a woman who was
Gabrielle Todd et al.
PloS one, 8(2), e56438-e56438 (2013-02-19)
Use of illicit stimulants such as methamphetamine, cocaine, and ecstasy is an increasing health problem. Chronic use can cause neurotoxicity in animals and humans but the long-term consequences are not well understood. The aim of the current study was to
J Burucker et al.
Fortschritte der Neurologie-Psychiatrie, 80(12), 684-693 (2012-12-12)
Substance-related disorders are clinically and socially very important. In Germany over a million people of varying ages are affected. Depending on the substance and stage of treatment, drugs and different treatment methods are used. Through a literature search we examined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service