Skip to Content
Merck
All Photos(1)

Key Documents

M28200

Sigma-Aldrich

1-(Methylamino)anthraquinone

≥97.5% purity, powder or crystals

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$133.00
100 G
$386.00
500 G
$1,490.00

$133.00


Estimated to ship onMay 10, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$133.00
100 G
$386.00
500 G
$1,490.00

About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$133.00


Estimated to ship onMay 10, 2025


Request a Bulk Order

Product Name

1-(Methylamino)anthraquinone, 97.5% (HPLC)

Quality Level

Assay

97.5% (HPLC)

form

powder or crystals

color

faint red to very dark red

mp

170-172 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CNc1cccc2C(=O)c3ccccc3C(=O)c12

InChI

1S/C15H11NO2/c1-16-12-8-4-7-11-13(12)15(18)10-6-3-2-5-9(10)14(11)17/h2-8,16H,1H3

InChI key

SVTDYSXXLJYUTM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Lahikainen et al.
Nature communications, 9(1), 4148-4148 (2018-10-10)
A reconfigurable actuator is a stimuli-responsive structure that can be programmed to adapt different shapes under identical stimulus. Reconfigurable actuators that function without control circuitry and are fueled remotely are in great demand to devise adaptive soft robotic devices. Yet, obtaining
T C Marrs et al.
Human toxicology, 3(4), 289-308 (1984-08-01)
The repeated-dose inhalation toxicity of a smoke containing a mixture of three dyes was tested in female mice, rats and guinea pigs. The green component dye (Solvent Green 3) was retained in the lungs. Particularly in the rats marked collections

Questions

  1. Which solvent can be used to reconstitute item M28200, 1-(Methylamino)anthraquinone?

    1 answer
    1. It can be dissolved in acetone, ethanol, ethylene glycol ether, and linseed oil. It is slightly soluble in benzene and carbon tetrachloride, but insoluble in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service