Skip to Content
Merck
All Photos(1)

Key Documents

L0702000

Lisinopril dihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lisinopril, (S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

lisinopril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].NCCCC[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C21H31N3O5.2H2O/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15;;/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29);2*1H2/t16-,17-,18-;;/m0../s1

InChI key

CZRQXSDBMCMPNJ-ZUIPZQNBSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lisinopril dihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sarah J Trenfield et al.
International journal of pharmaceutics, 577, 119066-119066 (2020-01-27)
Three-dimensional printing (3DP) is a revolutionary technology in pharmaceuticals, enabling the personalisation of flexible-dose drug products and 3D printed polypills (polyprintlets). A major barrier to entry of this technology is the lack of non-destructive quality control methods capable of verifying
Zhongchao Ma et al.
Human cell, 33(2), 330-336 (2020-01-19)
Renal fibrosis is a common characteristic of chronic kidney disease (CKD), and it can lead to end-stage renal disease. It has been reported that silibinin or lisinopril (MK-521) can inhibit the progression of renal fibrosis. However, the effect of combination
Manal Eid et al.
Journal of separation science, 40(18), 3646-3654 (2017-07-20)
We present an environmentally friendly method for the analysis of three angiotensin-converting enzyme inhibitors and hydrochlorothiazide simultaneously using a green micellar eluent for the first time. The chromatographic separation of enalapril maleate, lisinopril dihydrate, benazepril hydrochloride, and hydrochlorothiazide was implemented
C Valenzuela et al.
General pharmacology, 25(5), 825-832 (1994-09-01)
1. The effects of lisinopril, an angiotensin-converting enzyme inhibitor, were studied on cardiac contractile force, action potential characteristics and membrane ionic currents. 2. In guinea-pig atria, lisinopril (0.001-1 microM) exerted a negative inotropic effect which was accompanied by a shortening
M C Nzerue
The Central African journal of medicine, 38(9), 391-392 (1992-09-01)
A case of angioedema induced by antihypertensive therapy with lisinopril is presented. The patient was a 70 year old black woman, with a history of hypertension for 15 years. The patient presented with acute onset of swelling involving the oro-facial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service