Keratin is typically insoluble. This product is not tested for amino acid content or purity.
Recommended Products
Product Name
Keratin azure, suitable for use as a protease substrate
Quality Level
form
solid
color
blue to very dark blue
suitability
suitable for protease substrate
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
1) Is this product soluble? 2) Is this product free of loose amino acids? How pure is it?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service