Skip to Content
Merck
All Photos(1)

Key Documents

IRMM653

Zinc 67Zn solution (certified for isotope abundance ratio)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,200.00

$1,200.00


Estimated to ship onMarch 21, 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$1,200.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,200.00


Estimated to ship onMarch 21, 2025


Request a Bulk Order

grade

certified reference material

Agency

IRMM®

isotopic purity

 67Zn

manufacturer/tradename

JRC

format

single component solution

Analysis Note

For more information please see:
IRMM653

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service