Skip to Content
Merck
All Photos(1)

Key Documents

I0775010

Isosorbide mononitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4:3,6-Dianhydro-D-glucitol 5-nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO6
CAS Number:
Molecular Weight:
191.14
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

isosorbide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])O[C@H]1[C@H]2OC[C@@H]([C@H]2OC1)O

InChI

1S/C6H9NO6/c8-3-1-11-6-4(13-7(9)10)2-12-5(3)6/h3-6,8H,1-2H2/t3-,4+,5+,6+/m0/s1

InChI key

YWXYYJSYQOXTPL-SLPGGIOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isosorbide mononitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayeed Hasan et al.
Journal of pharmaceutical sciences, 106(12), 3556-3563 (2017-08-27)
Extracellular polymeric substances in bacterial biofilms reduce the penetration of antimicrobials and give rise to extreme recalcitrance and treatment challenges for many persistent biofilms and associated infections. Nitric oxide (NO) is a promising alternative to conventional antimicrobials but is challenging
Tiantian Liu et al.
Chinese medicine, 15, 3-3 (2020-01-16)
The Chinese medicine Huoxue Wentong Formula (HXWTF) was used to treat thoracic obstruction and angina pectoris in clinic, which has not been investigated in myocardial ischemia-induced apoptosis and angiogenic function. Here we aimed to investigate the roles of HXWTF in
Francesca Fabrizi et al.
Journal of enzyme inhibition and medicinal chemistry, 27(1), 138-147 (2011-08-06)
The clinically used sulfonamide carbonic anhydrase (CA, EC 4.2.1.1) inhibitor dorzolamide (DRZ), a new sulfonamide CA inhibitor also incorporating NO-donating moieties, NCX250, and isosorbide mononitrate (ISMN) (an NO-donating compound with no CA inhibitory properties) were investigated for their intraocular pressure
Athanasios Mousiolis et al.
Contraception, 88(1), 41-44 (2013-01-08)
Isosorbide mononitrate (IMN) administration is thought to enhance in the cervical ripening process. We sought to identify the clinical usefulness and safety of IMN administration for shortening the time to termination of the second trimester pregnancy procedures when used in
U W Abshagen
The American journal of cardiology, 70(17), 61G-66G (1992-11-27)
Pharmacokinetic studies show that isosorbide mononitrate is rapidly absorbed after oral administration, reaches peak concentrations within an hour, undergoes no significant first-pass metabolism, and is virtually 100% bioavailable. The half-life is approximately 5 hours, the volume of distribution is 0.62

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service