Skip to Content
Merck
All Photos(1)

Documents

H9645

Supelco

p-Hydroxymethamphetamine

analytical standard, for drug analysis

Synonym(s):

Pholderin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO
CAS Number:
Molecular Weight:
165.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥98% (TLC)

drug control

Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CNC(C)Cc1ccc(O)cc1

InChI

1S/C10H15NO/c1-8(11-2)7-9-3-5-10(12)6-4-9/h3-6,8,11-12H,7H2,1-2H3

InChI key

SBUQZKJEOOQSBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A metabolite of methamphetamine; sympathomimetic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Yamamoto et al.
Journal of analytical toxicology, 13(2), 117-119 (1989-03-01)
A sensitive and specific gas chromatography/mass spectrometry (GC/MS) method has been developed for the simultaneous analysis of methampthetamine and its metabolites (amphetamine, p-hydroxymethamphetamine, and p-hydroxyamphetamine) in monkey urine. The method has been applied to the analysis of urine samples from
Ferenc Bánhidy et al.
Congenital anomalies, 50(2), 122-128 (2010-02-27)
Pholedrine was a frequently used drug for the treatment of severe hypotension in some countries, including Hungary. The possible teratogenic effect of pholedrine was not checked; therefore; the birth outcomes, particularly congenital abnormalities (CAs), of infants born to women treated
Rika Nakajima et al.
Chemical & pharmaceutical bulletin, 54(4), 493-495 (2006-04-06)
High-performance liquid chromatography (HPLC) with UV detection for the simultaneous determination of the free form of p-hydroxymethamphetamine (p-OHMA) and its metabolite, glucuronide (p-OHMAG) was accomplished for the first time. We achieved this by employing 1) an ion pair reagent for
N Kuroda et al.
Journal of chromatography. A, 798(1-2), 325-334 (1998-05-02)
Methods for the simultaneous determination of methamphetamine (MP) and its related compounds (MPs) using capillary electrophoresis (CE) with UV absorbance and laser-induced fluorescence (LIF) detection are described. In UV detection, MPs were applied to CE without any derivatization procedure and
N Shima et al.
Xenobiotica; the fate of foreign compounds in biological systems, 36(2-3), 259-267 (2006-05-17)
The urinary concentrations of the main metabolites of methamphetamine (MA), specifically p-hydroxymethamphetamine-sulfate (p-OHMA-Sul) and p-hydroxymethamphetamine-glucuronide (p-OHMA-Glu), were directly measured in MA users and rats using an optimized LC-ESI MS method. The concentrations of the two conjugates in 50 MA human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service