Skip to Content
Merck
All Photos(1)

Key Documents

H3766

Supelco

Sodium 4-hydroxybenzoate

analytical standard

Synonym(s):

Sodium p-hydroxybenzoate, 4-Hydroxybenzoic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$119.00

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$119.00

About This Item

Linear Formula:
4-HOC6H4CO2Na
CAS Number:
Molecular Weight:
160.10
Beilstein:
4163553
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$119.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[Na+].Oc1ccc(cc1)C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-3-1-5(2-4-6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ZLVSYODPTJZFMK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sodium 4-hydroxybenzoate has been used to study its catalytic esterification with benzyl bromide by ultrasound-assisted solid–liquid phase-transfer catalysis (U-SLPTC) using the novel dual-site phase-transfer catalyst 4,40-bis(tributylammoniomethyl)-1,10-biphenyl dichloride (BTBAMBC).[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jillian M Hagel et al.
Journal of industrial microbiology & biotechnology, 46(1), 91-99 (2018-11-06)
Since the 1930s, parabens have been employed widely as preservatives in food, pharmaceutical, and personal care products. These alkyl esters of benzoic acid occur naturally in a broad range of plant species, where they are thought to enhance overall fitness
Hung-Ming Yang et al.
Ultrasonics sonochemistry, 21(1), 395-400 (2013-08-27)
The catalytic esterification of sodium 4-hydroxybenzoate with benzyl bromide by ultrasound-assisted solid-liquid phase-transfer catalysis (U-SLPTC) was investigated using the novel dual-site phase-transfer catalyst 4,4'-bis(tributylammoniomethyl)-1,1'-biphenyl dichloride (BTBAMBC), which was synthesized from the reaction of 4,4'-bis(chloromethyl)-1,1'-biphenyl and tributylamine. Without catalyst and in
Ruth Pietri et al.
The Journal of biological chemistry, 287(28), 23748-23756 (2012-05-12)
Rhodopseudomonas palustris metabolizes aromatic compounds derived from lignin degradation products and has the potential for bioremediation of xenobiotic compounds. We recently identified four possible solute-binding proteins in R. palustris that demonstrated binding to aromatic lignin monomers. Characterization of these proteins
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 106, 175-184 (2013-02-06)
Large size and high quality single crystals of organic nonlinear optical material Piperidinium p-Hydroxybenzoate (PDPHB) have been grown by solution growth method. This crystal belongs to monoclinic system with a noncentrosymmetric space group of Cc. To confirm its structure and
Jian-Shuang Jiang et al.
Journal of Asian natural products research, 15(5), 427-432 (2013-04-23)
Two new glycoside compounds, named saffloquinoside C (1) and (-)-4-hydroxybenzoic acid-4-O-[6'-O-(2″-methylbutyryl)-β-D-glucopyranoside] (2), were isolated from the florets of Carthamus tinctorius. Their structures were elucidated by detailed spectroscopic means including UV, IR, HR-ESI-MS, 1D and 2D NMR, and CD data. Compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service