Key Documents
GX308088107
Supelco
Quartz Torch
for Elan/NexION
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
quality
for Elan/NexION
manufacturer/tradename
(30-808-8107)
compatibility
PerkinElmer 500, 5000, 6000, 9000
PerkinElmer Elan 6100DRC, DRC II, DRC-e
PerkinElmer NexION 300
General description
Choose GAZ-06 GazFit Connectors for easy connection of gas tubes to torch side arms.
Choose from one of the most recent versions:
Certificates of Analysis (COA)