Skip to Content
Merck
All Photos(1)

Key Documents

G6895

Supelco

Glafenine

analytical standard

Synonym(s):

1-Glyceryl N-(7-chloro-4-quinolyl)anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN2O4
CAS Number:
Molecular Weight:
372.80
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

OCC(O)COC(=O)c1ccccc1Nc2ccnc3cc(Cl)ccc23

InChI

1S/C19H17ClN2O4/c20-12-5-6-14-17(7-8-21-18(14)9-12)22-16-4-2-1-3-15(16)19(25)26-11-13(24)10-23/h1-9,13,23-24H,10-11H2,(H,21,22)

InChI key

GWOFUCIGLDBNKM-UHFFFAOYSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renaud Robert et al.
Molecular pharmacology, 77(6), 922-930 (2010-03-05)
Cystic fibrosis (CF) is caused by mutations in the CF transmembrane conductance regulator (CFTR) gene, which encodes a cAMP-activated anion channel expressed in epithelial cells. The most common mutation Delta Phe508 leads to protein misfolding, retention by the endoplasmic reticulum
C Moesch et al.
European journal of clinical chemistry and clinical biochemistry : journal of the Forum of European Clinical Chemistry Societies, 31(5), 329-333 (1993-05-01)
Glaphenine is an analgesic drug derived from anthranilic acid. We report the analytical procedures for stone analysis in three cases of common bile duct stones containing glaphenic acid, which developed in arthrosic patients treated for 13.7 +/- 5 years with
Bo Wen et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(9), 1511-1521 (2011-06-02)
Glafenine (Privadol; 2,3-dihydroxypropyl 2-[(7-chloro-4-quinolinyl) amino]benzoate) is a non-narcotic analgesic agent widely used for the treatment of pains of various origins. Severe liver toxicity and a high incidence of anaphylaxis were reported in patients treated with glafenine, eventually leading to its
A Davido et al.
La Revue de medecine interne, 10(2), 113-117 (1989-03-01)
Glafenine-induced shock. Seven cases. Glafenine is a widely prescribed analgesic drug, and shock is one of its severe side-effects. We report six documented and one highly probable cases of such accidents. From these seven cases and a review of the
Mariusz Swiader et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 12(4), 311-319 (2002-07-20)
It was previously documented that calcium (Ca(2+)) channel inhibitors intensified the protective effects of conventional antiepileptics against electroconvulsions in mice. The aim of this study was to evaluate the effects of Ca(2+) channel inhibitors (nifedipine, nicardipine and flunarizine) on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service