Skip to Content
Merck
All Photos(1)

Key Documents

FASTWORX-S

Sigma-Aldrich

FastWoRX-S Sorbent Powder

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$124.00

$124.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$124.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

$124.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Related Categories

Application

FastWoRX products are hydrophobic powders that absorb most organic compounds. With them, liquid-liquid extraction (LLE) for reaction work-ups can be replaced by a simple filtration or magnetic separation. This version of the product, FastWoRX-S, uses filtration. Work-ups can be done in half the time or less. Solvent use can be reduced by 90% or more. Emulsions cannot form. Automation of FastWoRX-based work-ups is much easier than automating LLE. For more details about the product, please see the attached flyer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

After filtration, the FastWoRX powder loaded with organic products can serve as a convenient input for flash chromatographic separation or other processing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service