Skip to Content
Merck

Skip To

ERMFA015

Charpy specimens 80 J (impact toughness)

ERM®, certified reference material

Synonym(s):

Charpy specimens (impact toughness)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

ERM®, certified by the BSC

manufacturer/tradename

JRC

application(s)

life science and biopharma

Packaging

set with 5 bars

Analysis Note

For more information please see:
ERMFA015

Legal Information

ERM is a registered trademark of European Commission


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
ERMFA015-1EA04061832660301

Questions

Reviews

No rating value

Active Filters