Skip to Content
Merck
All Photos(1)

Documents

ERMBF422B

Cotton seed GMO standard ERMBF422 for 281-24-236 x 3006-210-23

ERM®, certified reference material, level 1 (98% 281-24-236x3006-210-23 cotton seed), for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

quality

for the detection of GMO cotton seed (281-24-236x3006-210-23) with PCR

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBF422B

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service