About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95%
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
2-8°C
SMILES string
CCOc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CCN3C[C@@H]4CC)c2c1
InChI
1S/C21H28N2O2/c1-3-14-13-23-10-8-15(14)11-20(23)21(24)17-7-9-22-19-6-5-16(25-4-2)12-18(17)19/h5-7,9,12,14-15,20-21,24H,3-4,8,10-11,13H2,1-2H3/t14-,15-,20-,21+/m0/s1
InChI key
SUWZHLCNFQWNPE-LATRNWQMSA-N
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service