E0300000
Ephedrine hydrochloride
Drug Precursor, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(1R,2S)-(−)-Ephedrine hydrochloride, (1R,2S)-α-[1-(Methylamino)ethyl]benzyl alcohol hydrochloride, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol hydrochloride
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
ephedrine
manufacturer/tradename
EDQM
mp
216-220 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Cl.CN[C@@H](C)[C@H](O)c1ccccc1
InChI
1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m0./s1
InChI key
BALXUFOVQVENIU-GNAZCLTHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service