Skip to Content
Merck
All Photos(3)

Key Documents

E009755

Sigma-Aldrich

Pyridoxine hydrochloride

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

form

powder

color

white

mp

214-215 °C (lit.)

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin B6 is a water-soluble vitamin. It exists in six forms namely, pyridoxal, pyridoxine, pyridoxamine, pyridoxamine 5′-phosphate, pyridoxine 5′-phosphate and pyridoxal 5′-phosphate.[1] Pyridoxine is a pyrimidine derivative of vitamin B6. It is considered heat and acid stable.[2]

Biochem/physiol Actions

Vitamin B6 is an essential coenzyme for amino acid, carbohydrate and lipid metabolism. Pyridoxine serves as a primary source in multivitamins.[3] Pyridoxine deficiency is associated with peripheral neuropathy.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin B6 Deficiency (Pyridoxine)
StatPearls [Internet] (2018)
Revisiting the evidence for neuropathy caused by pyridoxine deficiency and excess
Ghavanini AA and Kimpinski K
Journal of Clinical Neuromuscular Disease, 16(1), 25-31 (2014)
Vitamin B6 (Pyridoxine; Pyridoxal 5'-Phosphate)
CH, C
Alternative Medicine Review, 6(1), 87-87 (2001)
Determination of vitamin B6 (pyridoxine) in pharmaceutical preparations by cyclic voltammetry at a copper (II) hexacyanoferrate (III) modified carbon paste electrode
Teixeira MFS, et al.
Journal of the Brazilian Chemical Society, 14(2), 316-321 (2003)
Vijay P Singh et al.
The Journal of organic chemistry, 78(4), 1478-1487 (2013-01-16)
Vitamin B6 is involved in a variety of enzymatic transformations. Some recent findings also indicate an antioxidant role of the vitamin in biological systems. We set out to turn pyridoxine (1a) into a catalytic chain-breaking and hydroperoxide-decomposing antioxidant by replacing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service