Skip to Content
Merck
All Photos(2)

Key Documents

D8787

Sigma-Aldrich

Dextran sulfate sodium salt from Leuconostoc spp.

avg mol wt >500,000 (dextran starting material)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$521.00

$521.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$521.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

$521.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

biological source

bacterial (Leuconostoc)

Quality Level

form

powder or crystals

mol wt

avg mol wt >500,000 (dextran starting material)

composition

Sulfur Content, 16.0-19.0% (dry basis)

color

white to off-white

pH

6.0-8.0

solubility

H2O: 100 mg/mL

Application


  • Determining the protective effects of Ma-Mu-Ran Antidiarrheal Capsules against acute DSS-induced enteritis using 16S rRNA gene sequencing and fecal metabolomics.: This study explores the protective effects of Ma-Mu-Ran on acute enteritis, highlighting the role of microbial interactions facilitated by dextran sulfate sodium salt, potentially beneficial for biochemists focusing on gut microbiome dynamics (Zheng et al., 2022).

  • Immunomodulatory and anti-inflammatory properties of artesunate in experimental colitis.: Examines the immunomodulatory effects of artesunate, potentially enhanced by adjuvants such as dextran sulfate sodium salt, pertinent to biochemists studying inflammation and immune responses (Yang et al., 2012).

  • Enantiomeric separation of drugs by affinity electrokinetic chromatography using dextran sulfate.: Investigates the application of dextran sulfate sodium salt in the chiral separation of pharmaceuticals, offering a technique of interest for biochemical analysis and drug development (Nishi et al., 1994).

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siroon Bekkering et al.
Clinical science (London, England : 1979), 133(10), 1185-1196 (2019-05-16)
Atherosclerosis is a chronic inflammatory disease that has its origins in early life. Postnatal inflammation exacerbates atherosclerosis, but the possible effect of intrauterine inflammation is largely unexplored. Exposure to inflammation in utero is common, especially in infants born preterm, who
Aaron H Rose et al.
Inflammatory bowel diseases, 21(9), 2005-2015 (2015-06-16)
An important role has emerged for calpain enzymes in regulating inflammation with one isoform, calpain-2, particularly important for macrophage activation. The goal of this study was to determine the therapeutic potential of a synthetic calpain-2 inhibitor, zLLY-CH2F, for colitis and

Protocols

Dextran sulfates as sodium salts are water-soluble and stable, containing approximately 17% sulfur for various applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service