Skip to Content
Merck
All Photos(1)

Key Documents

D7809

Millipore

Deoxycholate citrate agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$372.00

$372.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$372.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$372.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.5±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

selective and differential for Salmonella spp.
selective and differential for Shigella spp.

Application

A selective medium recommended for the isolation of enteric pathogens particularly Salmonella and Shigella species.

Components

Components (g/L)
Heart infusion solids 10.00
Proteose peptone 10.00
Lactose 10.00
Sodium citrate 20.00
Ferric ammonim citrate 2.00
Sodium deoxycholate 5.00
Neutral red 0.02
Agar 13.50

Preparation Note

Suspend 70.52 g of Deoxycholate Citrate Agar in 1000 ml of distilled water. Boil to dissolve the medium completely. Do not autoclave. Avoid excessive heating as it is detrimental to the medium.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The isolation of salmonellas and campylobacters.
C R Fricker
The Journal of applied bacteriology, 63(2), 99-116 (1987-08-01)

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service