Skip to Content
Merck

Skip To

D5000

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 5738 mPa.s (25 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mL
Check Cart for Availability
$241.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

$241.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

5738 mPa.s(25 °C)

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N350N2S3
format

mixture

format

mixture

format

mixture

format

mixture

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

viscosity

5738 mPa.s(25 °C)

viscosity

717.4 mPa.s(25 °C)

viscosity

2.144 mPa.s(25 °C)

viscosity

3.291 mPa.s(25 °C)


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters