D2935
Decarboxylase broth base, Moeller
suitable for microbiology, NutriSelect® Plus
About This Item
Recommended Products
sterility
non-sterile
Quality Level
product line
BioChemika
form
powder
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbe id | specific enzyme detection: suitable
microbiological culture: suitable
final pH
6.0±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
nonselective and differential for Citrobacter spp.
nonselective and differential for Enterobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Serratia spp.
nonselective and differential for Shigella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae
Application
Components
Peptic digest of animal tissue, 5.00
Beef extract, 5.00
Dextrose, 0.50
Bromocresol purple, 0.01
Cresol red, 0.005
Pyridoxal, 0.005
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service