Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 20 g | Check Cart for Availability | $224.00 |
About This Item
grade
certified reference material
Quality Segment
CofA
current certificate can be downloaded
packaging
pkg of 20 g
manufacturer/tradename
RTC CRM017-020
technique(s)
AAS: suitable, ICP: suitable
pH
7.48
application(s)
environmental
format
matrix material
storage temp.
2-30°C
General description
- N.I.S.T. Traceable
1 of 1
This Item | |||
|---|---|---|---|
| application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
| technique(s) AAS: suitable, ICP: suitable | technique(s) AAS: suitable, ICP: suitable | technique(s) AAS: suitable, ICP: suitable | technique(s) AAS: suitable, ICP: suitable |
| format matrix material | format matrix material | format matrix material | format matrix material |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| pH 7.48 | pH 6.35 | pH - | pH - |
Analyte
- Lead
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Repr. 1A - STOT RE 2
target_organs
Blood,Central nervous system,Immune system,Kidney
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| CRM017-20G | 04061834353546 |

