CAN43215
Residual Solvents, California Mix Categories I and II
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.21
Recommended Products
grade
certified reference material
Quality Level
CofA
current certificate can be downloaded
packaging
package of 1.2 ml
format
mixture
storage temp.
2-30°C
Components
- Analyte CAS Concentration
- Acetone 67-64-1 5000 µg/mL
- Acetonitrile 75-05-8 5000 µg/mL
- Ethanol 64-17-5 5000 µg/mL
- Ethyl acetate 141-78-6 5000 µg/mL
- Ethyl ether 60-29-7 5000 µg/mL
- Heptane 142-82-5 5000 µg/mL
- Hexane 110-54-3 5000 µg/mL
- Isopropyl alcohol 67-63-0 5000 µg/mL
- Methanol 67-56-1 5000 µg/mL
- Pentane 109-66-0 5000 µg/mL
- Toluene 108-88-3 5000 µg/mL
- m-xylene 108-38-3 5000 µg/mL
- o-xylene 95-47-6 5000 µg/mL
- p-xylene 106-42-3 5000 µg/mL
- Trichloroethylene 79-01-6 100 µg/mL
- 1,2-Dichloroethane 107-06-2 100 µg/mL
- Methylene Chloride 75-09-2 100 µg/mL
- Chloroform 67-66-3 100 µg/mL
- Benzene 71-43-2 100 µg/mL
- Dimethylacetamide 127-19-5 Solvent/Matrix
Analyte
Description
Acetonitrile 5000 ug/mL
Acetone 5000 ug/mL
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1A - Eye Irrit. 2 - Muta. 1B - Repr. 1B
WGK
WGK 3
Flash Point(F)
147.2 °F
Flash Point(C)
64 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service