Skip to Content
Merck
All Photos(1)

Key Documents

C8397

Supelco

Cysteamine S-phosphate sodium salt

analytical standard

Synonym(s):

S-(2-Aminoethyl) thiophosphate monosodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$219.00

$219.00


Estimated to ship onApril 29, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$219.00

About This Item

Linear Formula:
C2H7NO3PSNa
CAS Number:
Molecular Weight:
179.11
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$219.00


Estimated to ship onApril 29, 2025


Request a Bulk Order

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[Na+].NCCSP(O)([O-])=O

InChI

1S/C2H8NO3PS.Na/c3-1-2-8-7(4,5)6;/h1-3H2,(H2,4,5,6);/q;+1/p-1

InChI key

VZEGNYNPDZDIRW-UHFFFAOYSA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic hydrolysis of S-phosphocysteamine.
G Orzeszko et al.
Acta poloniae pharmaceutica, 45(2), 171-175 (1988-01-01)
[Toxicity and radiation-protective effect of multi-component combinations of sulfur-containing substances].
G V Kalistratov et al.
Radiobiologiia, 22(5), 628-632 (1982-09-01)
Andrea Annibal et al.
Nature communications, 12(1), 3486-3486 (2021-06-11)
The metabolome represents a complex network of biological events that reflects the physiologic state of the organism in health and disease. Additionally, specific metabolites and metabolic signaling pathways have been shown to modulate animal ageing, but whether there are convergent
[The effect of cystaphos on survival in combined radiation injuries].
M Simović
Vojnosanitetski pregled, 41(1), 3-11 (1984-01-01)
[Efficacy of cystaphos in protecting animals from gamma irradiation].
A Milovanović et al.
Vojnosanitetski pregled, 40(5), 325-328 (1983-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service