Skip to Content
Merck
All Photos(1)

Documents

C6272

Supelco

Clomiphene citrate salt

analytical standard

Synonym(s):

2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28ClNO · C6H8O7
CAS Number:
Molecular Weight:
598.08
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOc1ccc(cc1)\C(=C(\Cl)c2ccccc2)c3ccccc3

InChI

1S/C26H28ClNO.C6H8O7/c1-3-28(4-2)19-20-29-24-17-15-22(16-18-24)25(21-11-7-5-8-12-21)26(27)23-13-9-6-10-14-23;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,3-4,19-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25+;

InChI key

PYTMYKVIJXPNBD-BTKVJIOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clomiphene is an semisynthetic antiestrogen, widely used in the treatment of infertility by inducing ovulation and stimulating sperm production. It is also used in the treatment of breast cancer.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Mixture of cis and trans isomers

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Maryam Ardalan et al.
Pediatric neurosurgery, 48(3), 152-156 (2013-02-23)
Craniosynostosis is a relatively common congenital abnormality. The underlying etiology and most probable risk factors of nonsyndromic craniosynostosis are unknown. We conducted a study to identify the risk factors for craniosynostosis. In this case-control project, 70 children with craniosynostosis (syndromic
DNA adduct formation by tamoxifen and structurally-related compounds in rat liver
Rajaniemi H, et al.
Chemico-Biological Interactions, 113(2), 145-159 (1998)
Li Tan et al.
Journal of alternative and complementary medicine (New York, N.Y.), 18(12), 1087-1100 (2012-12-04)
The aim of this systematic review is to assess the effectiveness and safety of Chinese herbal medicine (CHM) in treatment of anovulation and infertility in women. Eight (8) databases were extensively retrieved. The Chinese electronic databases included VIP Information, CMCC
Jennifer Hesemann et al.
Spine, 38(5), E293-E298 (2013-03-01)
A retrospective medical record review of cases with congenital vertebral malformations (CVMs) and controls with normal spine morphology. To determine the relative contribution of maternal environmental factors (MEFs) during pregnancy to CVM development. CVMs represent defects in formation and segmentation
Polycystic ovarian syndrome.
Vanessa Yawn
Advance for NPs & PAs, 3(12), 11-14 (2012-12-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service