Skip to Content
Merck
All Photos(1)

Key Documents

C60

Flow Cup Viscosity Standard

UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 135.9 mPa.s (20 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$234.00

$234.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$234.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$234.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

135.9 mPa.s(20 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Flow Cup Viscosity Standards can be found on the Paragon Scientific webpage

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Passet et al.
Scientific reports, 10(1), 6765-6765 (2020-04-23)
Shadoo belongs to the prion protein family, an evolutionary conserved and extensively studied family due to the implication of PrP in Transmissible Spongiform Encephalopathies. However, the biological function of these genes remains poorly understood. While Sprn-knockdown experiments suggested an involvement
Carla Rocha-Santos et al.
Frontiers in physiology, 11, 309-309 (2020-04-25)
Aerobic exercise training improves the autonomic control of the circulation. Emerging evidence has shown that exercise induces neuroplastic adaptive changes in preautonomic circuitry controlling sympathetic/parasympathetic outflow to heart and vessels. The mechanisms underlying neuronal plasticity are, however, incompletely understood. Knowing
Milena D Vukic et al.
EXCLI journal, 19, 48-70 (2020-02-11)
In the present work we modified the procedure for isolation of naphthoquinones α-methylbutyrylshikon ( 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service