Skip to Content
Merck
All Photos(2)

Key Documents

C3561

Sigma-Aldrich

Pyrocatechol

suitable for plant cell culture, BioReagent, ≥99%, powder

Synonym(s):

1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
471401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 75 °C)
10 mmHg ( 118.3 °C)

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | plant: suitable

color

white to faint beige

bp

245 °C (lit.)

mp

100-103 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

Oc1ccccc1O

InChI

1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H

InChI key

YCIMNLLNPGFGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

May discolor on exposure to air and light.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christian J Kastrup et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(52), 21444-21449 (2012-12-14)
The treatment of diseased vasculature remains challenging, in part because of the difficulty in implanting drug-eluting devices without subjecting vessels to damaging mechanical forces. Implanting materials using adhesive forces could overcome this challenge, but materials have previously not been shown
Myung-Hyun Ryou et al.
Advanced materials (Deerfield Beach, Fla.), 25(11), 1571-1576 (2013-01-03)
Conjugation of mussel-inspired catechol groups to various polymer backbones results in materials suitable as silicon anode binders. The unique wetness-resistant adhesion provided by the catechol groups allows the silicon nanoparticle electrodes to maintain their structure throughout the repeated volume expansion
R D Irons
Journal of toxicology and environmental health, 16(5), 673-678 (1985-01-01)
Occupational exposure to benzene has long been associated with toxicity to the blood and bone marrow, including lymphocytopenia, pancytopenia, aplastic anemia, acute myelogenous leukemia, and possibly lymphoma. A variety of studies have established that benzene itself is not the toxic
R Zand et al.
Clinical pharmacology and therapeutics, 54(2), 142-149 (1993-08-01)
We studied the effect of isoniazid administration on the cytochrome P4502E1-catalyzed elimination of chlorzoxazone and acetaminophen. Isoniazid, 300 mg daily, was administered for 7 days to a group of 10 volunteer slow acetylators. Acetaminophen, 500 mg, and chlorzoxazone, 750 mg
Jacques Poupaert et al.
Current medicinal chemistry, 12(7), 877-885 (2005-04-28)
The 2(3H)-benzoxazolone heterocycle and its bioisosteric surrogates (such as 2(3H)-benzothiazolinone, benzoxazinone, etc.) have received considerable attention from the medicinal chemists owing to their capacity to mimic a phenol or a catechol moiety in a metabolically stable template. These heterocycles and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service