Skip to Content
Merck
All Photos(1)

Key Documents

C3298

Supelco

Casanthranol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C21H22O10/c22-6-8-4-10-15(12(25)5-8)17(27)14-9(2-1-3-11(14)24)20(10)31-21-19(29)18(28)16(26)13(7-23)30-21/h1-5,13,16,18-26,28-29H,6-7H2/t13-,16+,18-,19-,20-,21+/m1/s1

InChI key

CPUHNROBVJNNPW-VVBPCJSVSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Quality

Total hydroxyanthracene derivatives ≥20%, calculated as cascaroside A; ≥80% of the total hydroxyanthracene derivatives are cascarosides, calculated as cascaroside A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service