Skip to Content
Merck
All Photos(1)

Key Documents

C2040

Supelco

Cloperastine hydrochloride

analytical standard

Synonym(s):

4-Chlorobenzhydryl 2-(1-piperidyl)ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClNO · HCl
CAS Number:
Molecular Weight:
366.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.Clc1ccc(cc1)C(OCCN2CCCCC2)c3ccccc3

InChI

1S/C20H24ClNO.ClH/c21-19-11-9-18(10-12-19)20(17-7-3-1-4-8-17)23-16-15-22-13-5-2-6-14-22;/h1,3-4,7-12,20H,2,5-6,13-16H2;1H

InChI key

UNPLRYRWJLTVAE-UHFFFAOYSA-N

Application

Cloperastine hydrochloride may be used as an analytical standard for the quantification of cloperastine in biological samples using liquid chromatography coupled to tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Effect of cloperastine on muco-ciliary clearance].
D Olivieri et al.
Archivio Monaldi per la tisiologia e le malattie dell'apparato respiratorio, 38(5-6), 219-223 (1983-09-01)
[Cloperastine fendizoate in the treatment of cough-producing diseases in pediatrics].
L Scotti et al.
Minerva pediatrica, 40(5), 283-286 (1988-05-01)
Hao Li et al.
Journal of pharmaceutical and biomedical analysis, 51(3), 716-722 (2009-11-03)
A rapid and sensitive method based on liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the simultaneous quantitation of paracetamol, caffeine, pseudoephedrine, chlorpheniramine and cloperastine in human plasma has been developed and validated. After sample preparation by liquid-liquid extraction, the analytes and
Yukoh Sakata et al.
International journal of pharmaceutics, 335(1-2), 12-19 (2006-11-23)
Tablets containing both paracetamol (PM) and cloperastine hydrochloride (CLH) in a combination formulation prepared by standard vertical granulation technology were found to have altered pharmaceutical properties. The hardness and disintegration time of tablets containing both PM and CLH gradually increased
Gen Yamamoto et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 131(4), 523-532 (2011-04-07)
Clinically, both overactive bladder (OAB) and dysuria are known to occur in patients with cerebral infarction (CI). A few anticholinergic drugs are used to treat OAB in such patients, although the effect is not satisfactory. On the other hand, little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service